2-Deoxy-D-galactose 99%
Leverantör: Thermo Scientific
111970010 111970050
ACRO111970010EA 1300 SEK
ACRO111970010 ACRO111970050
2-Deoxy-D-galactose 99%
2-Deoxy-D-galactose
CAS No.: 1949-89-9
Formel: C₆H₁₂O₅
MW: 164,16 g/mol
Smältpunkt: 109…111 °C
Storage Temperature: Omgivande
MDL Number: MFCD00014649
CAS nummer: 1949-89-9
Order Now
Specifikation testresultat
Appearance White to light beige Crystalline powder or crystals
Infrared spectrum Conforms
HPLC ≥98.5 % (ELSD)
Specific optical rotation +56° to +60° (20°C, 589 nm) (c=2, H2O, 5 h)