N,N-Dimethyl-1,4-phenylenediamineoxalate 99%
Leverantör: Thermo Scientific
Fara
Synonyms: N,N-Dimethyl-p-phenylenediamine oxalate, 4-Aminodimethylaniline Oxalate
408490250 408490050 408491000
ACRO408490250EA 2525 SEK
ACRO408490250 ACRO408490050 ACRO408491000
N,N-Dimethyl-1,4-phenylenediamineoxalate 99%
N,N-Dimethyl-1,4-phenylenediamineoxalate
CAS No.: 62778-12-5
Formel: C₈H₁₂N₂
MW: 362,43 g/mol
Smältpunkt: 200 °C
Storage Temperature: Omgivande
MDL Number: MFCD00036387
CAS nummer: 62778-12-5
UN: 2811
ADR: 6.1,II
Order Now
Specifikation testresultat
Appearance Beige to grey to brown Powder and/or chunks
Infrared spectrum Conforms
Titration with NaOH ≥98.5 %
HPLC ≥97.0 %