4-(Dicyanomethylene)-2-methyl-6-(4-dimethylaminostyryl)-4H-pyran 96+%
Leverantör: Thermo Scientific
Varning
Synonyms: 2-(2-(4-(Dimethylamino)styryl)-6-methyl-4H-pyran-4-ylidene)malononitrile, DCM, 4-(Dicyanomethylene)-2-methyl-6-(p-dimethylaminostyryl)-4H-pyran
407072500 407070010
ACRO407072500EA 815 SEK
ACRO407072500 ACRO407070010
4-(Dicyanomethylene)-2-methyl-6-(4-dimethylaminostyryl)-4H-pyran 96+%
4-(Dicyanomethylene)-2-methyl-6-(4-dimethylaminostyryl)-4H-pyran
Appearance: Red to red-brown Powder or crystals
Formel: C₁₉H₁₇N₃O
MW: 303,37 g/mol
Smältpunkt: 212 °C
Flampunkt: 43 °C
Storage Temperature: Omgivande
MDL Number: MFCD00051341
CAS nummer: 51325-91-8
UN: 1325
ADR: 4.1,III
Order Now
Specifikation testresultat
Appearance Red to red-brown Powder or crystals
Infrared spectrum Conforms
HPLC ≥96.0 %
UV Lambda max 479 to 483 nm (in DMSO)
Molar absorption coefficient ≥42000 l/(mol.cm) (at Lambda max)