Bis(2-ethylhexyl)terephthalate 97%
Leverantör: Thermo Fisher Scientific
Synonyms: Bis(2-ethylhexyl) terephthalate
402490010 402492500 402490100
ACRO402490010EA 1025 SEK
ACRO402490010 ACRO402492500 ACRO402490100
Bis(2-ethylhexyl)terephthalate 97%
Bis(2-ethylhexyl)terephthalate
Appearance: Clear colorless Liquid
Formel: C₂₄H₃₈O₄
MW: 390.564 g/mol
Kokpunkt: 400 °C (760 mmHg)
Smältpunkt: –67 °C
Densitet: 0.984 g/cm³
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00072256
CAS nummer: 6422-86-2
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4870 to 1.4920 (20°C, 589 nm)
GC ≥96.0 %