Ethyl-5-chloromethyl-2-furancarboxylate 95%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 5-Chloromethyl-2-furancarboxylic Acid Ethyl Ester, Ethyl 5-(chloromethyl)furan-2-carboxylate
349730010 349730050 349730250
ACRO349730010EA 565 SEK
ACRO349730010 ACRO349730050 ACRO349730250
Ethyl-5-chloromethyl-2-furancarboxylate 95%
Ethyl-5-chloromethyl-2-furancarboxylate
CAS No.: 2528-00-9
Formel: C₈H₉ClO₃
MW: 188.61 g/mol
Kokpunkt: 115 °C (760 mmHg)
Densitet: 1.225 g/cm³
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00173837
CAS nummer: 2528-00-9
Order Now
Specifikation testresultat
Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5100 to 1.5130 (20°C, 589 nm)
GC ≥94 %