(Ethylthio)trimethylsilane 90%
Leverantör: Thermo Fisher Scientific
Fara
345000010 345000100
ACRO345000010EA 635 SEK
ACRO345000010 ACRO345000100
(Ethylthio)trimethylsilane 90%
(Ethylthio)trimethylsilane
Appearance: Clear colorless Liquid
Formel: C₅H₁₄SSi
Kokpunkt: 69…70 °C (100 mmHg)
Densitet: 0,83 g/cm³
Flampunkt: 16 °C
Storage Temperature: Omgivande
MDL Number: MFCD00042890
CAS nummer: 5573-62-6
UN: 1993
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4470 to 1.4510 (20°C, 589 nm)
GC ≥88 %