6-Hydroxyhexanoic acid 95%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 6-Hydroxycaproic acid
342240010 342240050 342240250
ACRO342240010EA 1025 SEK
ACRO342240010 ACRO342240050 ACRO342240250
6-Hydroxyhexanoic acid 95%
6-Hydroxyhexanoic acid
CAS No.: 1191-25-9
Formel: C₆H₁₂O₃
Kokpunkt: 113…116 °C (760 mmHg)
Smältpunkt: 38…40 °C
Densitet: 0,981 g/cm³
Storage Temperature: Kylskåp
MDL Number: MFCD00046560
CAS nummer: 1191-25-9
Order Now
Specifikation testresultat
Appearance White Low melting crystalline solid
Infrared spectrum Conforms
Additional info may contain variable amounts of water
GC ≥94 %