Trimethyl(vinyloxy)silane 97%
Leverantör: Thermo Scientific
|
Fara
|
338700010
338700050
ACRO338700010EA
605
SEK
ACRO338700010
ACRO338700050
Trimethyl(vinyloxy)silane 97%
Trimethyl(vinyloxy)silane
Appearance: Clear light yellow Liquid
Formel:
C₅H₁₂OSi MW: 116,24 g/mol Kokpunkt: 74…75 °C (760 mmHg) Densitet: 0,77 g/cm³ Flampunkt: –10 °C Storage Temperature: Kylskåp |
MDL Number:
MFCD00054764 CAS nummer: 6213-94-1 UN: 1993 ADR: 3,II |
Specifikation testresultat
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3885 to 1.3905 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...