cis-2-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid 98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: cis-2-(Boc-amino)cyclohexanecarboxylic acid
334270010 334270050
ACRO334270010EA 2025 SEK
ACRO334270010 ACRO334270050
cis-2-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid 98%
cis-2-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid
CAS No.: 63216-49-9
Formel: C₁₂H₂₁NO₄
MW: 243.30 g/mol
Smältpunkt: 127…131 °C
Flampunkt: 194 °C
Storage Temperature: Omgivande
MDL Number: MFCD01863244
CAS nummer: 63216-49-9
Order Now
Specifikation testresultat
Appearance White Powder
Infrared spectrum Conforms
Melting point 127 to 131 °C
Titration with NaOH ≥97.5 %