(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminocobalt(II) 98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: (1R,2R)-(-)-1,2-Cyclohexanediamino-N,N'-bis(3,5-di-t-butylsalicylidene)cobalt (II), (R,R)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminocobalt(II), (R,R)-Jacobsen's catalyst cobalt(II) complex
329950010 329950050 329950250
ACRO329950010EA 1150 SEK
ACRO329950010 ACRO329950050 ACRO329950250
(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminocobalt(II) 98%
(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminocobalt(II)
CAS No.: 176763-62-5
Formel: C₃₆H₅₄CoN₂O₂
MW: 603.74 g/mol
Smältpunkt: >350 °C
Storage Temperature: Omgivande
MDL Number: MFCD01631277
CAS nummer: 176763-62-5
Order Now
Specifikation testresultat
Appearance Orange to red-brown Powder
Infrared spectrum Conforms
Elemental Analysis ≥97.5 %