DETA NONOate (NOC-18) 95%
Leverantör: Thermo Scientific
Varning
Synonyms: DETA/NO, 2,2′-(Hydroxynitrosohydrazino)bis-ethanamine, Diethylenetriamine NONOate, 3,3-Bis(aminoethyl)-1-hydroxy-2-oxo-1-triazene
328650250 328651000
ACRO328650250EA 1300 SEK
ACRO328650250 ACRO328651000
DETA NONOate (NOC-18) 95%
DETA NONOate (NOC-18)
CAS No.: 146724-94-9
Formel: C₄H₁₃N₅O₂
MW: 163,181 g/mol
Smältpunkt: 98…106 °C
Storage Temperature: Frys
MDL Number: MFCD00278808
CAS nummer: 146724-94-9
Order Now
Specifikation testresultat
Appearance White to off-white Crystalline solid
Infrared spectrum Conforms
NMR ≥94 %