Wortmannin 98%
Leverantör: Thermo Scientific
Fara
Synonyms: (1S,6bR,9aS,11R,11bR)-1-(Methoxymethyl)-9a,11b-dimethyl-3,6,9-trioxo-1,6,6b,7,8,9,9a,10,11,11b-decahydro-3H-furo[4,3,2-de]indeno[4,5-h]isochromen-11-yl acetate
328590010 328590050
ACRO328590010EA 1975 SEK
ACRO328590010 ACRO328590050
Wortmannin 98%
Wortmannin
Appearance: White to light yellow Powder
Formel: C₂₃H₂₄O₈
MW: 428,44 g/mol
Smältpunkt: 234…240 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00133927
CAS nummer: 19545-26-7
UN: 2811
ADR: 6.1,I
Order Now
Specifikation testresultat
Appearance White to light yellow Powder or solid or film
HPLC ≥97.5 %