2-(Benzyloxy)ethanol 98%
Leverantör: Thermo Scientific
Varning
Synonyms: Ethylene Glycol Monobenzyl Ether, Benzyl Cellosolve, Benzyl Glycol
314650050 314650010 314650250
ACRO314650050EA 1675 SEK
ACRO314650050 ACRO314650010 ACRO314650250
2-(Benzyloxy)ethanol 98%
2-(Benzyloxy)ethanol
CAS No.: 622-08-2
Formel: C₉H₁₂O₂
MW: 152,19 g/mol
Kokpunkt: 265 °C
Densitet: 1,07 g/cm³
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002868
CAS nummer: 622-08-2
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5200 to 1.5220 (20°C, 589 nm)
GC ≥97.5 %