Ethoxycarbonyl isocyanate 90% tech.
Leverantör: Thermo Fisher Scientific
Varning
314310010 314310050 314310250
ACRO314310010EA 2475 SEK
ACRO314310010 ACRO314310050 ACRO314310250
Ethoxycarbonyl isocyanate 90% tech.
Ethoxycarbonyl isocyanate
CAS No.: 19617-43-7
Formel: C₄H₅NO₃
Kokpunkt: 25 °C (10 mmHg)
Densitet: 1,11 g/cm³
Flampunkt: 35 °C
Storage Temperature: Omgivande
MDL Number: MFCD00011719
CAS nummer: 19617-43-7
UN: 3080
ADR: 6.1,II
Order Now
Specifikation testresultat
Appearance Clear colorless to slightly colored Liquid
Infrared spectrum Conforms
GC ≥88 %