Ethoxyacetylene 40% (w/w) i hexaner
Leverantör: Thermo Scientific
Fara
Synonyms: ethynoxyethane, Ethyl ethynyl ether, ethoxy-ethyne
307390050 307390010 307390250
ACRO307390050EA 2200 SEK
ACRO307390050 ACRO307390010 ACRO307390250
Ethoxyacetylene 40% (w/w) i hexaner
Ethoxyacetylene
CAS No.: 927-80-0
Formel: C₄H₆O
MW: 70,091 g/mol
Kokpunkt: 55…56 °C (760 mmHg)
Densitet: 0,718 g/cm³
Flampunkt: –28 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00009247
CAS nummer: 927-80-0
UN: 1993
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Clear orange to brown Solution
Identification confirmed by 1H-NMR
NMR (1H-NMR) ca. 40 % w/w (Typical 35 to 55 % w/w)