1-Hydroxy-1-cyclopropanecarboxylic acid 97%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 1-hydroxycyclopropane-1-carboxylic acid, 1-Hydroxycyclopropanecarboxylic acid
301430010 301430050
ACRO301430010EA 2250 SEK
ACRO301430010 ACRO301430050
1-Hydroxy-1-cyclopropanecarboxylic acid 97%
1-Hydroxy-1-cyclopropanecarboxylic acid
CAS No.: 17994-25-1
Formel: C₄H₆O₃
MW: 102.089 g/mol
Smältpunkt: 108…110 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00010797
CAS nummer: 17994-25-1
UN: 3261
ADR: 8 III
Order Now
Specifikation testresultat
Appearance White to yellow to beige Powder
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %