4-Hydroxy-1,2,2,6,6-pentamethylpiperidine 99%
Leverantör: Thermo Scientific
Fara
Synonyms: 1,2,2,6,6-Pentamethylpiperidin-4-ol, 1,2,2,6,6-Pentamethyl-4-piperidinol
300860010 300862500 300860050
ACRO300860010EA 2800 SEK
ACRO300860010 ACRO300862500 ACRO300860050
4-Hydroxy-1,2,2,6,6-pentamethylpiperidine 99%
4-Hydroxy-1,2,2,6,6-pentamethylpiperidine
Appearance: Light yellow Flakes
Formel: C₁₀H₂₁NO
MW: 171,28 g/mol
Kokpunkt: 238 °C (760 mmHg)
Smältpunkt: 70…76 °C
Densitet: 0,967 g/cm³
Flampunkt: 118 °C
Storage Temperature: Omgivande
MDL Number: MFCD00191220
CAS nummer: 2403-89-6
UN: 3259
ADR: 8,II
Order Now
Specifikation testresultat
Appearance Light yellow Flakes
Infrared spectrum Conforms
Melting point 70 to 76 °C
GC ≥98.5 %