4-Hydroxy-7-methoxycoumarin 95%
Leverantör: Thermo Scientific
Varning
Synonyms: 4-Hydroxy-7-methoxy-2H-chromen-2-one
298850010 298850050
ACRO298850010EA 1600 SEK
ACRO298850010 ACRO298850050
4-Hydroxy-7-methoxycoumarin 95%
4-Hydroxy-7-methoxycoumarin
CAS No.: 17575-15-4
Formel: C₁₀H₈O₄
MW: 192,17 g/mol
Smältpunkt: 263…265 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00673700
CAS nummer: 17575-15-4
Order Now
Specifikation testresultat
Appearance Beige to pink Powder
Infrared spectrum Conforms
HPLC ≥94 %