cis,cis-Muconic acid 98%
Leverantör: Thermo Scientific
|
Varning
|
Synonyms:
(2Z,4Z)-Hexa-2,4-dienedioic acid
297760025
297760100
ACRO297760025EA
1550
SEK
ACRO297760025
ACRO297760100
cis,cis-Muconic acid 98%
cis,cis-Muconic acid
CAS No.: 1119-72-8
Formel:
C₆H₆O₄ MW: 142,11 g/mol Smältpunkt: 194…195 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00085266 CAS nummer: 1119-72-8 |
Specifikation testresultat
Appearance | Yellow to light beige Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...