N-Boc-3-(4-cyanophenyl)oxaziridine 98+%

Leverantör: Thermo Scientific
Varning

297095000 297090020
ACRO297095000EA 5200 SEK
ACRO297095000 ACRO297090020
N-Boc-3-(4-cyanophenyl)oxaziridine 98+%
N-Boc-3-(4-cyanophenyl)oxaziridine

CAS No.: 150884-56-3

Formel: C₁₃H₁₄N₂O₃
Smältpunkt: 57…63 °C
Storage Temperature: Frys
MDL Number: MFCD00800493
CAS nummer: 150884-56-3
UN: 3439
ADR: 6.1,III

Order Now

Specifikation testresultat

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 57 to 63 °C
Elemental Analysis ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR