4,6-Dimethyl-2-pyrone 98%
Leverantör: Thermo Scientific
295350100 295350010
ACRO295350100EA 3725 SEK
ACRO295350100 ACRO295350010
4,6-Dimethyl-2-pyrone 98%
4,6-Dimethyl-2-pyrone
CAS No.: 675-09-2
Formel: C₇H₈O₂
MW: 124,139 g/mol
Smältpunkt: 48…50 °C
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00075555
CAS nummer: 675-09-2
Order Now
Specifikation testresultat
Appearance yellow Low melting crystalline mass (May darken on storage)
Infrared spectrum Conforms
GC ≥97.5 %