Geranyl linalool mixture of isomers ∼95% tech.
Leverantör: Thermo Scientific
Varning
278360250 278360025
ACRO278360250EA 12625 SEK
ACRO278360250 ACRO278360025
Geranyl linalool mixture of isomers ∼95% tech.
Geranyl linalool
CAS No.: 1113-21-9
Formel: C₂₀H₃₄O
MW: 290,491 g/mol
Kokpunkt: 250 °C (760 mmHg)
Densitet: 0,88 g/cm³
Flampunkt: 160 °C
Storage Temperature: Omgivande
MDL Number: MFCD00059363
CAS nummer: 1113-21-9
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4860 to 1.4920 (20°C, 589 nm)
GC ≥94 %
Specific gravity (25°C) 0.875 to 0.885