diexo-3-Amino-bicyclo[2.2.1]hept-5-ene-2-carboxylic acid amide 99+%
Leverantör: Thermo Scientific
Synonyms: 3-exo-Aminobicyclo[2.2.1]hept-5-ene-2-exo-carboxamide
277812500 277810010
ACRO277812500EA 1675 SEK
ACRO277812500 ACRO277810010
diexo-3-Amino-bicyclo[2.2.1]hept-5-ene-2-carboxylic acid amide 99+%
diexo-3-Amino-bicyclo[2.2.1]hept-5-ene-2-carboxylic acid amide
CAS No.: 105786-40-1
Formel: C₈H₁₂N₂O
MW: 152,197 g/mol
Smältpunkt: 235…253 °C
Storage Temperature: Omgivande
MDL Number: MFCD00143157
CAS nummer: 105786-40-1
Order Now
Specifikation testresultat
Appearance White Crystalline powder
Infrared spectrum Conforms
GC ≥99.0 %