Jod lösning 0,1 mol ICl/l acc. Wijs
Leverantör: Thermo Scientific
Fara
Synonyms: Chloroiodide, Iodine monochloride, Iodochloride, Iodine chloride
270570010 270570025
ACRO270570010EA 1775 SEK
ACRO270570010 ACRO270570025
Jod lösning 0,1 mol ICl/l acc. Wijs
Jodklorid
Formel: ClI
MW: 162,36 g/mol
Smältpunkt: 17,2 °C
Densitet: 1,06 g/cm³
Flampunkt: 40 °C
Storage Temperature: Omgivande
MDL Number: MFCD00011354
CAS nummer: 7790-99-0
UN: 2920
ADR: 8,II
Order Now
Specifikation testresultat
Appearance Clear red-brown to brown Solution
Titration with Na2S2O3 0.098 to 0.113 M