Ethyl-3-(dimethylamino)acrylate 99+%
Leverantör: Thermo Scientific
Varning
Synonyms: Ethyl 3-(dimethylamino)acrylate, 3-(Dimethylamino)acrylic Acid Ethyl Ester, Ethyl 3-(N,N-dimethylamino)acrylate
269630250 269631000
ACRO269630250EA 1225 SEK
ACRO269630250 ACRO269631000
Ethyl-3-(dimethylamino)acrylate 99+%
Ethyl-3-(dimethylamino)acrylate
CAS No.: 924-99-2
Formel: C₇H₁₃NO₂
MW: 143,19 g/mol
Kokpunkt: 118…121 °C (10HPA)
Smältpunkt: 17…18 °C
Densitet: 1 g/cm³
Flampunkt: 105 °C
Storage Temperature: Omgivande
MDL Number: MFCD00144269
CAS nummer: 924-99-2
Order Now
Specifikation testresultat
Appearance Clear yellow to light orange Liquid
Infrared spectrum Conforms
Refractive index 1.5105 to 1.5125 (20°C, 589 nm)
GC ≥99.0 %