trans-2-Amino-1-cyclohexanecarboxylic acid 98%
Leverantör: Thermo Scientific
Varning
265630010 265630050
ACRO265630010EA 3925 SEK
ACRO265630010 ACRO265630050
trans-2-Amino-1-cyclohexanecarboxylic acid 98%
trans-2-Amino-1-cyclohexanecarboxylic acid
CAS No.: 5691-19-0
Formel: C₇H₁₃NO₂
MW: 143,186 g/mol
Smältpunkt: 274…278 °C
Storage Temperature: Omgivande
MDL Number: MFCD00145418
CAS nummer: 5691-19-0
Order Now
Specifikation testresultat
Appearance White to off-white Powder and/or chunks
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %