2-(Methyldithio)isobutyraldehyde 95%
Leverantör: Thermo Scientific
Varning
265150010 265150050
ACRO265150010EA 1375 SEK
ACRO265150010 ACRO265150050
2-(Methyldithio)isobutyraldehyde 95%
2-(Methyldithio)isobutyraldehyde
CAS No.: 67952-60-7
Formel: C₅H₁₀OS₂
Kokpunkt: 46 °C (1 mmHg)
Densitet: 1,1 g/cm³
Flampunkt: 72 °C
Storage Temperature: Omgivande
MDL Number: MFCD00142943
CAS nummer: 67952-60-7
Order Now
Specifikation testresultat
Appearance Clear light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5190 to 1.5230 (20°C, 589 nm)
GC ≥94 %