2-(4-Isobutylphenyl)propanoic acid 99%
Leverantör: Thermo Scientific
Varning
Synonyms: (±)-Ibuprofen, [(±)-2-(4-Isobutylphenyl)-propionic Acid, Ibuprofen
258610100 258610500 258610010
ACRO258610100EA 5375 SEK
ACRO258610100 ACRO258610500 ACRO258610010
2-(4-Isobutylphenyl)propanoic acid 99%
2-(4-Isobutylphenyl)propanoic acid
CAS No.: 15687-27-1
Formel: C₁₃H₁₈O₂
MW: 206,3 g/mol
Kokpunkt: 157 °C (4 mmHg)
Smältpunkt: 75…78 °C
Storage Temperature: Omgivande
MDL Number: MFCD00010393
CAS nummer: 15687-27-1
Order Now
Specifikation testresultat
Appearance White to almost white Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 75 to 78 °C
Titration with NaOH 98.5 to 101.5 %