Hydrogen peroxide-urea 35% H₂O₂/g (w/w) stabilised
Leverantör: Thermo Scientific
Fara
Synonyms: Urea hydrogen peroxide
241020010 241022500 241020051
ACRO241020010EA 2425 SEK
ACRO241020010 ACRO241022500 ACRO241020051
Hydrogen peroxide-urea 35% H₂O₂/g (w/w) stabilised
Hydrogen peroxide-urea
1 g tablets, contains 35 wt% H2O2
Formel: CH₆N₂O₃
MW: 94,07 g/mol
Smältpunkt: 75…85 °C (dec.)
Storage Temperature: Kylskåp
MDL Number: MFCD00013119
CAS nummer: 124-43-6
UN: 1511
ADR: 5.1,III
Order Now
Specifikation testresultat
Appearance White Tablets
Stabilizer 1 % Sodium pyrophosphate
Titration with KMnO4 ≥33.5 % (H2O2)