L-Buthionine sulphoximine 99%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: L-Buthionine-(S,R)-sulphoximine, (2S)-2-Amino-4-(butylsulphonimidoyl)butanoic acid
235520010 235520050
ACRO235520010EA 6225 SEK
ACRO235520010 ACRO235520050
L-Buthionine sulphoximine 99%
L-Buthionine sulphoximine
CAS No.: 83730-53-4
Formel: C₈H₁₈N₂O₃S
MW: 222.303 g/mol
Smältpunkt: 234…235 °C (dec.)
Storage Temperature: Kylskåp
MDL Number: MFCD00067000
CAS nummer: 83730-53-4
Order Now
Specifikation testresultat
Appearance White Powder
Infrared spectrum Conforms
TLC ≥98.5 %
Specific optical rotation +31° to +35° (20°C, 589 nm) (c=1, 1 N HCl)