Phytic acid 50% (w/w) i vattenlösning
Leverantör: Thermo Scientific
Fara
Synonyms: Inositol hexaphosphoric acid
235370010 235372500 235370025
ACRO235370010EA 4325 SEK
ACRO235370010 ACRO235372500 ACRO235370025
Phytic acid 50% (w/w) i vattenlösning
Phytic acid
CAS No.: 83-86-3
Formel: C₆H₁₈O₂₄P₆
MW: 660,03 g/mol
Kokpunkt: 105 °C (760 mmHg)
Densitet: 1,37 g/cm³
Storage Temperature: Omgivande
MDL Number: MFCD00082309
CAS nummer: 83-86-3
UN: 3265
ADR: 8,II
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Solution
Titration with NaOH 45 to 55 % w/w