Carbonylcyanide-3-chlorophenylhydrazone 98%
Leverantör: Thermo Scientific
Fara
Synonyms: CCCP, Carbonyl cyanide m-chlorophenylhydrazone, Carbonyl cyanide m-chlorophenylhydrazone
228130010 228131000
ACRO228130010EA 2550 SEK
ACRO228130010 ACRO228131000
Carbonylcyanide-3-chlorophenylhydrazone 98%
Carbonylcyanide-3-chlorophenylhydrazone
Appearance: Orange to orange-brown Crystalline powder
Formel: C₉H₅ClN₄
MW: 204,617 g/mol
Smältpunkt: 172…177 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00001848
CAS nummer: 555-60-2
UN: 2811
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance Yellow to orange to orange-brown Powder
Infrared spectrum Conforms
HPLC ≥97.5 %