N-α-Benzoyl-DL-arginine-4-nitroanilide hydrochloride 98%
Leverantör: Thermo Scientific
Synonyms: N-α-Benzoyl-DL-arginine-4-nitroanilide hydrochloride, BAPNA, BANI, Bz-DL-Arg-pNA·HCl
227740010 227740050
ACRO227740010EA 1550 SEK
ACRO227740010 ACRO227740050
N-α-Benzoyl-DL-arginine-4-nitroanilide hydrochloride 98%
N-α-Benzoyl-DL-arginine-4-nitroanilide hydrochloride
CAS No.: 911-77-3
Formel: C₁₉H₂₃ClN₆O₄
MW: 434,881 g/mol
Smältpunkt: 268…272 °C (dec.)
Storage Temperature: Kylskåp
MDL Number: MFCD00012846
CAS nummer: 911-77-3
Order Now
Specifikation testresultat
Appearance White to yellow to light brown Powder
Infrared spectrum Conforms
HPLC ≥97.5 %