6-Mercaptopurine monohydrate 99.5+%
Leverantör: Thermo Scientific
Varning
Synonyms: 6-Purinethiol monohydrate, 6-Mercaptopurine hydrate, 7H-Purine-6-thiol hydrate, 6-Thiohypoxanthine monohydrate
226520050 226520250
ACRO226520050EA 1425 SEK
ACRO226520050 ACRO226520250
6-Mercaptopurine monohydrate 99.5+%
6-Mercaptopurine monohydrate
Appearance: Yellow Powder
Formel: C₅H₄N₄S
MW: 170,19 g/mol (152,17 anhy)
Smältpunkt: 313 °C
Storage Temperature: Omgivande
MDL Number: MFCD01461928
CAS nummer: 6112-76-1
Order Now
Specifikation testresultat
Appearance Yellow Powder
Infrared spectrum Conforms
Water 9 to 13 % (K.F.)
HPLC ≥99.50 %