Carnauba wax, yellow, refined No. 1
Leverantör: Thermo Fisher Scientific
222380010 222382500 222380050
ACRO222380010EA 2125 SEK
ACRO222380010 ACRO222382500 ACRO222380050
Carnauba wax, yellow, refined No. 1
Carnauba wax
CAS No.: 8015-86-9
Smältpunkt: 81…86 °C
Flampunkt: 282 °C
Storage Temperature: Omgivande
MDL Number: MFCD00130724
CAS nummer: 8015-86-9
Order Now
Specifikation testresultat
Appearance Yellow Flakes
Melting point 80 to 88°C
Residue after ignition ≤0.25 %
Acid value 2 to 7 mg KOH/g
Saponification value 78 to 95 mg KOH/g