Kaliumbromat 1 N i vattenlösning USP Test Solution (TS) 34
Leverantör: VWR Chemicals
85334.180EA 1975 SEK
85334.180 85334.260
Kaliumbromat 1 N i vattenlösning USP Test Solution (TS) 34
Kaliumbromat
Ready to use test solutions (TS).
  • Produced according to Pharmacopoeia requirements
  • Reduces time and expenses
  • Independent, traceable and certified
  • Conforms to USP 34
  • Certificates of analysis and safety data sheets available
Formel: KBrO₃
MW: 167 g/mol
Smältpunkt: -7,2 °C
Storage Temperature: Omgivande
MDL Number: MFCD00011359
CAS nummer: 7758-01-2
EINECS: 0
ADR: 8,I
Merck Index: 13,01378