Kaliumpermanganat 0.1 N aqueous solution USP Test Solution (TS)
Leverantör: VWR Chemicals
Synonyms: Permanganic acid potassium salt
85367.180EA 385 SEK
85367.180 85367.260
Kaliumpermanganat 0.1 N aqueous solution USP Test Solution (TS)
Kaliumpermanganat
Ready to use test solution (TS).
  • Produced according to Pharmacopoeia requirements
  • Reduces time and expenses
  • Independent, traceable and certified
  • Conforms to USP 34
  • Certificates of analysis and safety data sheets available
Formel: KMnO₄
MW: 158.03 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00011364
CAS nummer: 7722-64-7
EINECS: 0
UN: 3082
ADR: 9 III
Merck Index: 12,07824