Ammoniumtiocyanat i vattenlösning USP Test Solution (TS)
Leverantör: VWR Chemicals
Synonyms: Thiocyanic acid ammonium salt, Ammonium rhodanide, Ammonium sulphocyanate
85331.180EA 640 SEK
85331.180 85331.260
Ammoniumtiocyanat i vattenlösning USP Test Solution (TS)
Ammoniumtiocyanat
Ready to use test solution (TS).
  • Produced according to Pharmacopoeia requirements
  • Reduces time and expenses
  • Independent, traceable and certified
  • Conforms to USP 34
  • Certificates of analysis and safety data sheets available
Formel: NH₄SCN
MW: 76.12 g/mol
Smältpunkt: 150
Storage Temperature: Omgivande
MDL Number: MFCD00011428
CAS nummer: 1762-95-4
EINECS: 0
Merck Index: 13,00564