Chromatography cartridges, Protein L, Pierce™
Leverantör: Thermo Fisher Scientific
89928 89929
PIER89928EA 5725 SEK
PIER89928 PIER89929
Chromatography cartridges, Protein L, Pierce™
Kromatografikolonn
Pierce™ Protein L chromatography cartridges are convenient, ready-to-use prepacked devices for isolation and purification of immunoglobulin classes IgG, IgM, IgA, IgE and IgD via their kappa light chains.
  • Immobilised Protein L is ideal for selective purification of human and mouse antibodies that have kappa light chains
  • Resin support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
  • Superior manufacturing method immobilises Protein L by charge-free, leach-resistant covalent bonds, resulting in low non specific binding and enabling multiple uses without decline in yield
  • Binding capacity of 5 to 10 mg human IgG/ml resin
Pierce™ Protein L chromatography cartridges are compatible with the major automated liquid-chromatography systems or manual syringe processing. The cartridges attach directly to ÄKTA™ or FPLC Systems without additional connectors. The included accessory pack readily adapts cartridges for use with Luer-Lok® syringe fittings or ¹/₁₆" tubing. These cartridges enable fast, easy and reproducible chromatographic separations and can be regenerated for multiple uses.
Leveransinformation: Kit includes an accessory pack (1 female Luer-Lok® adapter, 1 connector fitting, 1 column plug and bottom caps).