Alphazurine A, pulver C.I. 42080
Leverantör: MP Biomedicals
Synonyms: Fenazo bluexg, Patent Blue A, Carmine Blue AF, Alphazurine A, Erio Glaucine X, Acid Blue 7
0215473025 0215473080
ICNA0215473025EA 880 SEK
ICNA0215473025 ICNA0215473080
Alphazurine A, pulver C.I. 42080
Alphazurine A
Soluble in water (1 mg/mL).
Formel: C₃₇H₃₅N₂NaO₆S₂
MW: 690.805 g/mol
MDL Number: MFCD00044440
CAS nummer: 3486-30-4