Cellulose, microcrystalline NF
Leverantör: Spectrum Chemical
Synonyms: Microcrystallinecellulose
C1679-12KGBL C1679-2.5KG C1679-50KGBL
SPCMC1679-12KGBLEA 18275 SEK
SPCMC1679-12KGBL SPCMC1679-2.5KG SPCMC1679-50KGBL
Cellulose, microcrystalline NF
Cellulose
Allergen statement (available upon request)
Formel: (C₁₂H₂₂O₁₁)n
MW: 342.3 g/mol
Smältpunkt: 260…270 °C
Densitet: 1.27…1.6 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00081512
CAS nummer: 9004-34-6
EINECS: 232-674-9
Merck Index: 13,01977