L (+)-Cysteinhydrokloridmonohydrat USP
Leverantör: Spectrum Chemical
Synonyms: (R)-(+)-Cysteine hydrochloride monohydrate, (R)-2-Amino-3-mercaptopropanoic acid hydrochloride hydrate, L-Cysteine HCI Monohydrate, H-Cys-OH.HCl.H2O, Cysteine Hydrochloride, Monohydrate, Cysteine Hydrochloride monohydrate
C1473-12KGBL C1473-1KG C1473-50KGBL
SPCMC1473-12KGBLEA 0 SEK
SPCMC1473-12KGBL SPCMC1473-1KG SPCMC1473-50KGBL
L (+)-Cysteinhydrokloridmonohydrat USP
L (+)-Cysteinhydrokloridmonohydrat
BSE/TSE statement (available upon request)
Allergen statement (available upon request)
Formel: HSCH₂CH(NH₂)COOH·HCl·H₂O
MW: 175,64 g/mol
Smältpunkt: 168…170 °C
Storage Temperature: Omgivande
MDL Number: MFCD00065606
CAS nummer: 7048-04-6
EINECS: 200-157-7
Merck Index: 13,02810
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18703c57a6b5f299a5f38f5d92b333d3d65de497&vid=f5322d0e0e22206df920d9255aee728bf83dd711&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Assay 98.5% Min.