Benzylbenzoat USP
Leverantör: Spectrum Chemical
Varning
Synonyms: Benzoic acid benzyl ester
BE159-200LTBL BE159-20LT
SPCMBE159-200LTBLEA 0 SEK
SPCMBE159-200LTBL SPCMBE159-20LT
Benzylbenzoat USP
Benzylbenzoat
BSE/TSE statement (available upon request)
Allergen statement (available upon request)
Formel: C₁₄H₁₂O₂
MW: 212,25 g/mol
Kokpunkt: 323 °C (1013 hPa)
Smältpunkt: 17…20 °C
Densitet: 1,118 g/cm³ (20 °C)
Flampunkt: 158 °C (closed cup)
Storage Temperature: Omgivande
MDL Number: MFCD00003075
CAS nummer: 120-51-4
EINECS: 204-402-9
UN: 3082
ADR: 9,III
Merck Index: 13,01129
Order Now
Specifikation testresultat
Assay 99% Min.