Vitamin A palmitate USP
Leverantör: Spectrum Chemical
Varning
Synonyms: Retinyl palmitate, Retinol palmitate, all-trans-Retinyl palmitate
V1159-1LT V1159-5KGBL
SPCMV1159-1LTEA 36550 SEK
SPCMV1159-1LT SPCMV1159-5KGBL
Vitamin A palmitate USP
Vitamin A palmitate
BSE/TSE statement (available upon request)
Allergen statement (available upon request)
Formel: C₃₆H₆₀O₂
MW: 524,87 g/mol
MDL Number: MFCD00019414
CAS nummer: 79-81-2
EINECS: 201-228-5
Order Now
Specifikation testresultat
Assay 95% Min.