Undec-10-enoic acid USP
Leverantör: Spectrum Chemical
Varning
Synonyms: Undecylenic-10 acid, Undecenoic acid, 10-Undecenoic acid, Undecylenic acid, Undecenylic acid
UN105-20LT UN105-4LT
SPCMUN105-20LTEA 0 SEK
SPCMUN105-20LT SPCMUN105-4LT
Undec-10-enoic acid USP
Undec-10-enoic acid
Allergen statement (available upon request)
Formel: C₁₁H₂₀O₂
MW: 184.28 g/mol
Kokpunkt: 300
Smältpunkt: 24
Densitet: 0.91 g/cm³ (20 °C)
Flampunkt: 146
MDL Number: MFCD00004442
CAS nummer: 112-38-9
EINECS: 203-965-8
Merck Index: 12,09983
Order Now
Specifikation testresultat
Assay 97% Min.