Trietanolamin NF
Leverantör: Spectrum Chemical
Synonyms: Tris-(2-hydroxyethyl)amine, 2,2',2''-Nitrilotriethanol
TR143-200LTBL TR143-20LT TR143-4LTGL
SPCMTR143-200LTBLEA 90350 SEK
SPCMTR143-200LTBL SPCMTR143-20LT SPCMTR143-4LTGL
Trietanolamin NF
Trietanolamin
BSE/TSE statement (available upon request)
Allergen statement (available upon request)
Formel: C₆H₁₅NO₃
MW: 149,19 g/mol
Kokpunkt: 335,4 °C (1013 hPa)
Smältpunkt: 21,6 °C
Densitet: 1,13 g/cm³ (20 °C)
Flampunkt: 179 °C (closed cup)
Storage Temperature: Omgivande
MDL Number: MFCD00002855
CAS nummer: 102-71-6
EINECS: 203-049-8
Merck Index: 13,09739
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c181d3e80375aec7c34e17766247484e1c3d3257&vid=53d40d0e56c8683dfbafebfbeba65d9447c1bb55&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Assay 99% Min.