Tiaminhydroklorid USP
Leverantör: Spectrum Chemical
Synonyms: Aneurine hydrochloride, 3-((4-Amino-2-methylpyrimidin-5-yl)methyl)-5-(2-hydroxyethyl)-4-methylthiazol-3-ium chloride hydrochloride, Thiaminium chloride, VitamineB1 hydrochloride
TH116-1KG TH116-25KGBL TH116-25GM TH116-500GM
SPCMTH116-1KGEA 11875 SEK
SPCMTH116-1KG SPCMTH116-25KGBL SPCMTH116-25GM SPCMTH116-500GM
Tiaminhydroklorid USP
Tiaminhydroklorid
Formel: C₁₂H₁₈Cl₂N₄OS
MW: 337.27 g/mol
Smältpunkt: 252
Densitet: 1.401 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00012780
CAS nummer: 67-03-8
EINECS: 200-641-8
Merck Index: 13,09366
Order Now
Specifikation testresultat
Assay 98% Min.