D-(+)-Sucrose octaacetate NF
Leverantör: Spectrum Chemical
Synonyms: D-(+)-Saccharose octaacetate, Sucrose octaacetate, D(+)-Sucroseoctaacetat, (2R,3R,4S,5R,6R)-2-(Acetoxymethyl)-6-(((2S,3S,4R,5R)-3,4-diacetoxy-2,5-bis(acetoxymethyl)tetrahydrofuran-2-yl)oxy)tetrahydro-2H-pyran-3,4,5-triyl triacetate
SU104-12KG SU104-2.5KG SU104-25KGBL SU104-50KGBL
SPCMSU104-12KGEA 35150 SEK
SPCMSU104-12KG SPCMSU104-2.5KG SPCMSU104-25KGBL SPCMSU104-50KGBL
D-(+)-Sucrose octaacetate NF
D-(+)-Sucrose octaacetate
BSE/TSE statement (available upon request)
Allergen statement (available upon request)
Formel: C₂₈H₃₈O₁₉
MW: 678,6 g/mol
Smältpunkt: 80…83 °C
Densitet: 1,257 g/cm³ (20 °C)
Storage Temperature: Kylskåp
MDL Number: MFCD00006623
CAS nummer: 126-14-7
EINECS: 204-772-1
Merck Index: 12,09052
Order Now
Specifikation testresultat
Assay 98% Min.