L (+)-Lysin monohydroklorid, granular USP
Leverantör: Spectrum Chemical
Synonyms: Lysine Hydrochloride, (S)-(+)-2,6-Diaminohexanoic acid monohydrochloride, L-Lysine monohydrochloride, H-Lys-OH·HCl, L-Lysine monohydrocholoride, L-Lysine Hydrochloride, (S)-(+)-Lysine monohydrochloride, (S)-(+)-2,6-Diaminocaproic acid monohydrochloride, H-Lys-OH.HCl
L1142-1KG L1142-50KGBL L1142-5KG
SPCML1142-1KGEA 4250 SEK
SPCML1142-1KG SPCML1142-50KGBL SPCML1142-5KG
L (+)-Lysin monohydroklorid, granular USP
L (+)-Lysin monohydroklorid
Form: Granular
Use: excipient, biochemistry.
Formel: H₂N(CH₂)₄CH(NH₂)CO₂H·HCl
MW: 182,65 g/mol
Smältpunkt: 263…264 °C
Storage Temperature: Omgivande
MDL Number: MFCD00064564
CAS nummer: 657-27-2
EINECS: 211-519-9
Order Now
Specifikation testresultat
Assay 98.5% Min.