EDTA dikaliumsalt dihydrat ≥99%, Reagent Grade
Leverantör: Spectrum Chemical
Synonyms: Ethylenediaminetetraacetic acid dipotassium salt dihydrate, Titriplex® Potassium salt, Ethylenediaminetetra-acetic acid dipotassium salt dihydrate, Edetic acid dipotassium salt dihydrate, Dipotassium dihydrogen ethylenediaminetetraacetate dihydrate, Ethylenedinitrilotetra-acetic acid dipotassium salt dihydrate, EDTA dipotassium dihydrate, Potassium 2,2'-((2-(bis(carboxymethyl)amino)ethyl)azanediyl)diacetate dihydrate, Ethylenedinitrilotetraacetic acid dipotassium salt
E1042-12KG E1042-2.5KG E1042-50KGBL
SPCME1042-12KGEA 24925 SEK
SPCME1042-12KG SPCME1042-2.5KG SPCME1042-50KGBL
EDTA dikaliumsalt dihydrat ≥99%, Reagent Grade
EDTA dikaliumsalt dihydrat
Formel: C₁₀H₁₄K₂N₂O₈·2H₂O
MW: 404.46 g/mol
Smältpunkt: 272
Densitet: 1 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00150036
CAS nummer: 25102-12-9
EINECS: 217-895-0
Merck Index: 13,03546
Order Now
Specifikation testresultat
Assay 99% Min.